Product Details
- SNP ID
-
rs77458369
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:135558345 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATGTGGCTGGTAGCTACCTTATTA[A/G]ACAGCACAACTATCTAAATTTGGAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
R3HDM1
PubMed Links
Gene Details
- Gene
- R3HDM1
- Gene Name
- R3H domain containing 1
View Full Product Details