Product Details
- SNP ID
-
rs79015645
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:119157856 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCACTTCACCCTCGGAATCGCCACC[G/T]ACCCCGGCCCCGCCGCCCACCACCC
- Phenotype
-
MIM: 614330
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
C1QL2
PubMed Links
Gene Details
- Gene
- C1QL2
- Gene Name
- complement C1q like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182528.3 |
1040 |
Silent Mutation |
GTA,GTC |
V138V |
NP_872334.2 |
View Full Product Details