Product Details
- SNP ID
-
rs76204302
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:38298356 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGACCGCAAACAGTGTGGCTGGGG[G/T]ACGAGCAGCATAGAAGATATTTTTG
- Phenotype
-
MIM: 609368
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATL2
PubMed Links
Gene Details
- Gene
- ATL2
- Gene Name
- atlastin GTPase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001308076.1 |
1860 |
Missense Mutation |
ACC,CCC |
T456P |
NP_001295005.1 |
XM_011533017.1 |
1860 |
Missense Mutation |
ACC,CCC |
T474P |
XP_011531319.1 |
XM_011533018.2 |
1860 |
Missense Mutation |
ACC,CCC |
T469P |
XP_011531320.1 |
XM_011533019.2 |
1860 |
Missense Mutation |
ACC,CCC |
T456P |
XP_011531321.1 |
XM_011533020.1 |
1860 |
Missense Mutation |
ACC,CCC |
T420P |
XP_011531322.1 |
XM_011533022.2 |
1860 |
Missense Mutation |
ACC,CCC |
T303P |
XP_011531324.1 |
XM_011533023.2 |
1860 |
Missense Mutation |
ACC,CCC |
T247P |
XP_011531325.1 |
XM_017004686.1 |
1860 |
Missense Mutation |
ACC,CCC |
T420P |
XP_016860175.1 |
XM_017004687.1 |
1860 |
Missense Mutation |
ACC,CCC |
T420P |
XP_016860176.1 |
XM_017004688.1 |
1860 |
Missense Mutation |
ACC,CCC |
T247P |
XP_016860177.1 |
View Full Product Details