Product Details

SNP ID
rs76323369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:46621389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCTAGGAGTCTTCCGTTAGAG[A/C]TTACGTATCTGCCTCCTTCTTGCTG
Phenotype
MIM: 604594 MIM: 600153
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CRIPT PubMed Links

Gene Details

Gene
CRIPT
Gene Name
CXXC repeat containing interactor of PDZ3 domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014171.5 Intron NP_054890.1
Gene
PIGF
Gene Name
phosphatidylinositol glycan anchor biosynthesis class F
There are no transcripts associated with this gene.

View Full Product Details