Product Details

SNP ID
rs75513521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:53853257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAATCCAAAAGAGTTTACTGGCTG[G/T]AAGTTACATGGGCTGCTCTCCTCCT
Phenotype
MIM: 606704
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR75 PubMed Links

Gene Details

Gene
GPR75
Gene Name
G protein-coupled receptor 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006794.3 1815 Missense Mutation TTA,TTC L500F NP_006785.1
Gene
GPR75-ASB3
Gene Name
GPR75-ASB3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164165.1 1815 Intron NP_001157637.1
Gene
MIR3682
Gene Name
microRNA 3682
There are no transcripts associated with this gene.

View Full Product Details