Product Details

SNP ID
rs74422133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61871572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTACCATACAGACAATATAAAG[G/T]TGGACTGTCAGTTCTTTCCCATCAT
Phenotype
MIM: 605142
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCT4 PubMed Links

Gene Details

Gene
CCT4
Gene Name
chaperonin containing TCP1 subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256721.1 Intron NP_001243650.1
NM_006430.3 Intron NP_006421.2

View Full Product Details