Product Details

SNP ID
rs79299541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68489569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGAACACTGTTGGCTCAAAACAG[A/G]AAGCAAATAGATATTTTTAAAAAAT
Phenotype
MIM: 611035
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APLF PubMed Links

Gene Details

Gene
APLF
Gene Name
aprataxin and PNKP like factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173545.2 Intron NP_775816.1

View Full Product Details