Product Details
- SNP ID
-
rs75592412
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:68948708 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTGAGTCTTCACCTGGAGTGAAAA[C/T]TGTCTCTACAGAGTTCCAAATAATG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GKN2
PubMed Links
Gene Details
- Gene
- GKN2
- Gene Name
- gastrokine 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182536.2 |
|
Intron |
|
|
NP_872342.2 |
View Full Product Details