Product Details
- SNP ID
-
rs76235816
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:68950138 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTGCTGCTTACATGTTTATAGTC[A/C]AAAATTGTGGTAGAAGAGCATGATC
- Phenotype
-
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GKN2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs62133344] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GKN2
- Gene Name
- gastrokine 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182536.2 |
301 |
Missense Mutation |
TTG,TTT |
L64F |
NP_872342.2 |
View Full Product Details