Product Details

SNP ID
rs76527553
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26045896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAAAAAAAGAGTTTAAAGGATTA[C/T]AAAGGATGTTGAGGTTAACAGGACA
Phenotype
MIM: 612672
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RAB10 PubMed Links
Additional Information
For this assay, SNP(s) [rs113124685] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
RAB10
Gene Name
RAB10, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016131.4 Intron NP_057215.3

View Full Product Details