Product Details

SNP ID
rs74887298
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:26706228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCATCCTCGAGCCAAAGCTCCTG[A/G]GGTGAGCCAGGTGGCCAAGACATCA
Phenotype
MIM: 603220
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNK3 PubMed Links

Gene Details

Gene
KCNK3
Gene Name
potassium two pore domain channel subfamily K member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002246.2 Intron NP_002237.1
XM_005264293.2 Intron XP_005264350.1

View Full Product Details