Product Details

SNP ID
rs77205320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27131009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAAATGCCAGGGGCTTCATGT[C/G]AAGAGGAACTGGCCACAAGGCTGAG
Phenotype
MIM: 612195 MIM: 606395
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ABHD1 PubMed Links

Gene Details

Gene
ABHD1
Gene Name
abhydrolase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032604.3 1719 Intron NP_115993.3
XM_011533135.2 1719 Intron XP_011531437.1
XM_011533136.2 1719 Intron XP_011531438.1
XM_011533137.2 1719 Intron XP_011531439.1
XM_011533138.2 1719 Intron XP_011531440.1
XM_017005113.1 1719 Intron XP_016860602.1
Gene
PREB
Gene Name
prolactin regulatory element binding
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_006711914.2 1719 UTR 3 XP_006711977.1
XM_011532471.2 1719 UTR 3 XP_011530773.1
XM_011532472.2 1719 UTR 3 XP_011530774.1
Gene
PRR30
Gene Name
proline rich 30
There are no transcripts associated with this gene.

View Full Product Details