Product Details

SNP ID
rs80141585
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85606073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCAGTGACATCACCCATTGTGGC[C/T]GAGGAACCACAAAACCTTAAAATAC
Phenotype
MIM: 612490 MIM: 616757 MIM: 611594
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf68 PubMed Links

Gene Details

Gene
C2orf68
Gene Name
chromosome 2 open reading frame 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013649.3 3445 UTR 3 NP_001013671.2
XM_005264305.3 3445 Intron XP_005264362.1
Gene
RNF181
Gene Name
ring finger protein 181
There are no transcripts associated with this gene.

Gene
TMEM150A
Gene Name
transmembrane protein 150A
There are no transcripts associated with this gene.

Gene
USP39
Gene Name
ubiquitin specific peptidase 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256725.1 3445 Intron NP_001243654.1
NM_001256726.1 3445 Intron NP_001243655.1
NM_001256727.1 3445 Intron NP_001243656.1
NM_001256728.1 3445 Intron NP_001243657.1
NM_006590.3 3445 Intron NP_006581.2
XM_006711922.1 3445 Intron XP_006711985.1
XM_006711923.2 3445 Intron XP_006711986.1
XM_011532487.1 3445 Intron XP_011530789.1
XM_011532488.1 3445 Intron XP_011530790.1
XM_017003182.1 3445 Intron XP_016858671.1

View Full Product Details