Product Details
- SNP ID
-
rs76346420
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:28812469 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTGTGATTTACTTACCAAAAAA[A/G]AAAGAAAGAAAGAAAGAAAGAAAAT
- Phenotype
-
MIM: 600590
MIM: 614029
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PPP1CB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs541545678,rs555622461] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PPP1CB
- Gene Name
- protein phosphatase 1 catalytic subunit beta
There are no transcripts associated with this gene.
- Gene
- SPDYA
- Gene Name
- speedy/RINGO cell cycle regulator family member A
View Full Product Details