Product Details

SNP ID
rs75641430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:29098038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGAGGTGAGTATCCTGAACACA[A/G]CGTAGCATTATTTTAGACAGATGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLIP4 PubMed Links

Gene Details

Gene
CLIP4
Gene Name
CAP-Gly domain containing linker protein family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287527.1 Intron NP_001274456.1
NM_001287528.1 Intron NP_001274457.1
NM_024692.5 Intron NP_078968.3
XM_005264562.2 Intron XP_005264619.1
XM_005264563.1 Intron XP_005264620.1
XM_006712104.2 Intron XP_006712167.1
XM_006712105.3 Intron XP_006712168.1
XM_011533109.1 Intron XP_011531411.1
XM_011533110.1 Intron XP_011531412.1
XM_017004958.1 Intron XP_016860447.1
XM_017004959.1 Intron XP_016860448.1

View Full Product Details