Product Details

SNP ID
rs76734444
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:78601164 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTTGGCATGCTTTTCCCCACCT[A/C]CCCCAAATGCTTCCTCTACTATTCA
Phenotype
MIM: 602430
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ROBO1 PubMed Links

Gene Details

Gene
ROBO1
Gene Name
roundabout guidance receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145845.1 Intron NP_001139317.1
NM_002941.3 Intron NP_002932.1
NM_133631.3 Intron NP_598334.2
XM_006713277.2 Intron XP_006713340.1
XM_011533976.1 Intron XP_011532278.1
XM_011533977.2 Intron XP_011532279.1
XM_011533978.1 Intron XP_011532280.1
XM_011533979.1 Intron XP_011532281.1
XM_011533980.1 Intron XP_011532282.1
XM_017006982.1 Intron XP_016862471.1
XM_017006983.1 Intron XP_016862472.1
XM_017006984.1 Intron XP_016862473.1
XM_017006985.1 Intron XP_016862474.1

View Full Product Details