Product Details

SNP ID
rs77047020
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:10327348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTGTGACTACAGGAAGACAATTT[A/G]GTTTAAAATCACACTGAGTACATCT
Phenotype
MIM: 108733 MIM: 600152
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ATP2B2 PubMed Links

Gene Details

Gene
ATP2B2
Gene Name
ATPase plasma membrane Ca2+ transporting 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001331.2 5948 UTR 3 NP_001001331.1
NM_001683.3 5948 UTR 3 NP_001674.2
XM_005265179.4 5948 UTR 3 XP_005265236.1
XM_006713175.3 5948 UTR 3 XP_006713238.1
XM_011533752.2 5948 UTR 3 XP_011532054.1
XM_017006481.1 5948 UTR 3 XP_016861970.1
XM_017006482.1 5948 UTR 3 XP_016861971.1
XM_017006483.1 5948 UTR 3 XP_016861972.1
XM_017006484.1 5948 UTR 3 XP_016861973.1
XM_017006485.1 5948 UTR 3 XP_016861974.1
XM_017006486.1 5948 UTR 3 XP_016861975.1
XM_017006487.1 5948 UTR 3 XP_016861976.1
XM_017006488.1 5948 Intron XP_016861977.1
XM_017006489.1 5948 UTR 3 XP_016861978.1
XM_017006490.1 5948 Intron XP_016861979.1
XM_017006491.1 5948 UTR 3 XP_016861980.1
XM_017006492.1 5948 UTR 3 XP_016861981.1
Gene
MIR378B
Gene Name
microRNA 378b
There are no transcripts associated with this gene.

Gene
SEC13
Gene Name
SEC13 homolog, nuclear pore and COPII coat complex component
There are no transcripts associated with this gene.

View Full Product Details