Product Details

SNP ID
rs79069309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:24834456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTTTTGACAAAAAATAAAAATG[A/G]AAGCAAAATATTTGAGTGAGAAGTA
Phenotype
MIM: 180220
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RARB PubMed Links

Gene Details

Gene
RARB
Gene Name
retinoic acid receptor beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000965.4 Intron NP_000956.2
NM_001290216.2 Intron NP_001277145.1
NM_001290217.1 Intron NP_001277146.1
NM_001290266.1 Intron NP_001277195.1
NM_001290276.1 Intron NP_001277205.1
NM_001290277.1 Intron NP_001277206.1
NM_001290300.1 Intron NP_001277229.1
NM_016152.3 Intron NP_057236.1

View Full Product Details