Product Details

SNP ID
rs75816802
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:27716826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAATAAATACAGAACCTTGGATAC[C/T]CTTCGAATTTTAAAATACCTTAAAG
Phenotype
MIM: 604615
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EOMES PubMed Links

Gene Details

Gene
EOMES
Gene Name
eomesodermin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278182.1 2662 UTR 3 NP_001265111.1
NM_001278183.1 2662 UTR 3 NP_001265112.1
NM_005442.3 2662 UTR 3 NP_005433.2
XM_005265510.3 2662 UTR 3 XP_005265567.1

View Full Product Details