Product Details

SNP ID
rs79760825
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:28250574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTTCTTTCTGTAAATATTTATC[A/G]TGTTGCTTCTGTGTACTATATAACT
Phenotype
MIM: 615166
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CMC1 PubMed Links

Gene Details

Gene
CMC1
Gene Name
C-X9-C motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182523.1 Intron NP_872329.1
XM_011533402.2 Intron XP_011531704.1
XM_017005760.1 Intron XP_016861249.1

View Full Product Details