Product Details

SNP ID
rs78123694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39410335 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTTAAAAATTATGTAGTAAGAC[G/T]TTATTTCAATGTTGACAGGACGGGT
Phenotype
MIM: 150370 MIM: 180646
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RPSA PubMed Links

Gene Details

Gene
RPSA
Gene Name
ribosomal protein SA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304288.1 Intron NP_001291217.1
NM_002295.5 Intron NP_002286.2
Gene
SNORA6
Gene Name
small nucleolar RNA, H/ACA box 6
There are no transcripts associated with this gene.

Gene
SNORA62
Gene Name
small nucleolar RNA, H/ACA box 62
There are no transcripts associated with this gene.

View Full Product Details