Product Details

SNP ID
rs74549518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:40529896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTAAGTGAGCCCTTTGTTCCTTT[A/G]TCCATCCAGTTTTCTGGGGATGTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF621 PubMed Links

Gene Details

Gene
ZNF621
Gene Name
zinc finger protein 621
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098414.2 Intron NP_001091884.1
NM_001287245.1 Intron NP_001274174.1
NM_198484.4 Intron NP_940886.1
XM_005265079.3 Intron XP_005265136.1
XM_005265080.3 Intron XP_005265137.1
XM_006713114.3 Intron XP_006713177.1
XM_011533610.2 Intron XP_011531912.1
XM_017006226.1 Intron XP_016861715.1

View Full Product Details