Product Details

SNP ID
rs79098599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42017751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGTGAGAAATTTATTTAATTATT[A/G]TTGTTAATAGTGCACCTGAGGTGAA
Phenotype
MIM: 608112
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAK1 PubMed Links

Gene Details

Gene
TRAK1
Gene Name
trafficking kinesin protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042646.2 Intron NP_001036111.1
NM_001265608.1 Intron NP_001252537.1
NM_001265609.1 Intron NP_001252538.1
NM_001265610.1 Intron NP_001252539.1
NM_014965.4 Intron NP_055780.2
XM_005264956.2 Intron XP_005265013.1
XM_005264958.2 Intron XP_005265015.1
XM_005264960.3 Intron XP_005265017.1
XM_005264962.3 Intron XP_005265019.1
XM_005264963.3 Intron XP_005265020.1
XM_006713029.2 Intron XP_006713092.1
XM_006713030.2 Intron XP_006713093.1
XM_006713031.3 Intron XP_006713094.1
XM_011533489.2 Intron XP_011531791.1
XM_017005906.1 Intron XP_016861395.1
XM_017005907.1 Intron XP_016861396.1
XM_017005908.1 Intron XP_016861397.1
XM_017005909.1 Intron XP_016861398.1
XM_017005910.1 Intron XP_016861399.1
XM_017005911.1 Intron XP_016861400.1
XM_017005912.1 Intron XP_016861401.1
XM_017005913.1 Intron XP_016861402.1

View Full Product Details