Product Details

SNP ID
rs75188307
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42258191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTATCCTGTGGCTGGGGTCCAG[G/A]TTCTGCAGGTTCTTAACGATGGACA
Phenotype
MIM: 118440
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CCK PubMed Links

Gene Details

Gene
CCK
Gene Name
cholecystokinin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000729.5 1149 Silent Mutation AAC,AAT N85N NP_000720.1
NM_001174138.2 1149 Silent Mutation AAC,AAT N85N NP_001167609.1

View Full Product Details