Product Details

SNP ID
rs74843221
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45595204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGCTGCCAAGCCTCCTCTTGCT[A/G]CCTCGACAGGGGCACCTGGGGCAGT
Phenotype
MIM: 604543
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIMD1 PubMed Links

Gene Details

Gene
LIMD1
Gene Name
LIM domains containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014240.2 374 Missense Mutation ACC,GCC T109A NP_055055.1
XM_011534207.2 374 Missense Mutation ACC,GCC T109A XP_011532509.1

View Full Product Details