Product Details

SNP ID
rs75347427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45919705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTCTCCGCAGTATCCTTGGAAA[G/T]CCAAGCACTCAAAGATTCTAGATTG
Phenotype
MIM: 607182 MIM: 606568
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FYCO1 PubMed Links

Gene Details

Gene
FYCO1
Gene Name
FYVE and coiled-coil domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024513.3 6712 UTR 3 NP_078789.2
XM_006713333.3 6712 UTR 3 XP_006713396.1
XM_006713334.3 6712 UTR 3 XP_006713397.1
XM_011534111.2 6712 UTR 3 XP_011532413.1
Gene
LZTFL1
Gene Name
leucine zipper transcription factor like 1
There are no transcripts associated with this gene.

View Full Product Details