Product Details

SNP ID
rs78864109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48672523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAATGTTGCTGCTTCAACTTCT[A/G]AGAGCCTCTTGCTGGGGGGCCCCAT
Phenotype
MIM: 604264 MIM: 606671
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CELSR3 PubMed Links

Gene Details

Gene
CELSR3
Gene Name
cadherin EGF LAG seven-pass G-type receptor 3
There are no transcripts associated with this gene.

Gene
CELSR3-AS1
Gene Name
CELSR3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
NCKIPSD
Gene Name
NCK interacting protein with SH3 domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016453.3 Intron NP_057537.1
NM_184231.2 Intron NP_909119.1
XM_017006594.1 Intron XP_016862083.1
XM_017006595.1 Intron XP_016862084.1
XM_017006596.1 Intron XP_016862085.1
XM_017006597.1 Intron XP_016862086.1
XM_017006598.1 Intron XP_016862087.1
XM_017006599.1 Intron XP_016862088.1
XM_017006600.1 Intron XP_016862089.1

View Full Product Details