Product Details

SNP ID
rs76082653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48992035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGAAATAGATTATGCTGTCAGT[C/T]CTGTTGTGGGAGAGGCACCCAAAGC
Phenotype
MIM: 605615 MIM: 614584
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARIH2 PubMed Links

Gene Details

Gene
ARIH2
Gene Name
ariadne RBR E3 ubiquitin protein ligase 2
There are no transcripts associated with this gene.

Gene
P4HTM
Gene Name
prolyl 4-hydroxylase, transmembrane
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_177938.2 Intron NP_808807.2
NM_177939.2 Intron NP_808808.1

View Full Product Details