Product Details

SNP ID
rs75801519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50318173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGGGCTGCCAGAGCCAGCAGAC[C/T]GGTGAGGTGGGACCCAGCCCAGGCC
Phenotype
MIM: 607071 MIM: 603551 MIM: 607052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HYAL1 PubMed Links

Gene Details

Gene
HYAL1
Gene Name
hyaluronoglucosaminidase 1
There are no transcripts associated with this gene.

Gene
HYAL2
Gene Name
hyaluronoglucosaminidase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003773.4 1746 Missense Mutation AGT,GGT S460G NP_003764.3
NM_033158.4 1746 Missense Mutation AGT,GGT S460G NP_149348.2
XM_005265524.2 1746 Missense Mutation AGT,GGT S460G XP_005265581.1
XM_005265525.2 1746 Missense Mutation AGT,GGT S460G XP_005265582.1
Gene
TUSC2
Gene Name
tumor suppressor candidate 2
There are no transcripts associated with this gene.

View Full Product Details