Product Details

SNP ID
rs78491479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50348172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGGGGTGGCGGCCAATGAGGTCT[C/T]GCACGGTAGTGCCAGGGCTCAGGCT
Phenotype
MIM: 607068 MIM: 607072 MIM: 605082 MIM: 607069 MIM: 607070
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB561D2 PubMed Links

Gene Details

Gene
CYB561D2
Gene Name
cytochrome b561 family member D2
There are no transcripts associated with this gene.

Gene
NPRL2
Gene Name
NPR2-like, GATOR1 complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006545.4 1287 Missense Mutation CAA,CGA Q295R NP_006536.3
XM_005264808.4 1287 Missense Mutation CAA,CGA Q175R XP_005264865.1
XM_011533288.2 1287 Missense Mutation CAA,CGA Q292R XP_011531590.1
XM_017005555.1 1287 Missense Mutation CAA,CGA Q175R XP_016861044.1
XM_017005556.1 1287 Missense Mutation CAA,CGA Q175R XP_016861045.1
Gene
RASSF1
Gene Name
Ras association domain family member 1
There are no transcripts associated with this gene.

Gene
RASSF1-AS1
Gene Name
RASSF1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TMEM115
Gene Name
transmembrane protein 115
There are no transcripts associated with this gene.

Gene
ZMYND10
Gene Name
zinc finger MYND-type containing 10
There are no transcripts associated with this gene.

View Full Product Details