Product Details

SNP ID
rs77340141
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52267788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAAACAAACAAAAAACATGGTGG[C/T]GGTAGAGAAATGAAGTGCCATAAAG
Phenotype
MIM: 612102 MIM: 611059
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIRLET7G PubMed Links

Gene Details

Gene
MIRLET7G
Gene Name
microRNA let-7g
There are no transcripts associated with this gene.

Gene
WDR82
Gene Name
WD repeat domain 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025222.3 Intron NP_079498.2
XM_011534136.2 Intron XP_011532438.1

View Full Product Details