Product Details

SNP ID
rs75842407
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9813268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGAGGATGAAGATGAGGACGAG[A/G]AGTTCCAGCCATCACAGCTGTTCGA
Phenotype
MIM: 604226
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARPC4 PubMed Links

Gene Details

Gene
ARPC4
Gene Name
actin related protein 2/3 complex subunit 4
There are no transcripts associated with this gene.

Gene
ARPC4-TTLL3
Gene Name
ARPC4-TTLL3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198793.1 1094 Missense Mutation AAG,GAG K174E NP_001185722.1
Gene
TTLL3
Gene Name
tubulin tyrosine ligase like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025930.3 1094 Missense Mutation AAG,GAG K223E NP_001021100.3

View Full Product Details