Product Details

SNP ID
rs78359207
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:72032699 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACCAGAGTCCTTATTGAAGGCGT[C/T]TGGAGAGGTTTTATCCCCTTCTGGG
Phenotype
MIM: 607449
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NPFFR2 PubMed Links

Gene Details

Gene
NPFFR2
Gene Name
neuropeptide FF receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144756.1 Intron NP_001138228.1
NM_004885.2 Intron NP_004876.2
NM_053036.2 Intron NP_444264.1
XM_011531554.2 Intron XP_011529856.1

View Full Product Details