Product Details
- SNP ID
-
rs78574148
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:73406750 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGATGAGTCAGCTGAAAATTGT[A/G]ACAAATCACTTGTAAGTACATTCTA
- Phenotype
-
MIM: 103600
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ALB
PubMed Links
Gene Details
- Gene
- ALB
- Gene Name
- albumin
There are no transcripts associated with this gene.
View Full Product Details