Product Details

SNP ID
rs75165399
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:74145466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGGTGATGTCCTGGAGCCAGT[A/C]TCCCAGAATACCAAGGGATGGCTAC
Phenotype
MIM: 614047
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MTHFD2L PubMed Links

Gene Details

Gene
MTHFD2L
Gene Name
methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 2-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144978.1 Intron NP_001138450.1
XM_005265690.2 Intron XP_005265747.1
XM_017008218.1 Intron XP_016863707.1
XM_017008219.1 Intron XP_016863708.1
XM_017008220.1 Intron XP_016863709.1
XM_017008221.1 Intron XP_016863710.1
XM_017008222.1 Intron XP_016863711.1
XM_017008223.1 Intron XP_016863712.1
XM_017008224.1 Intron XP_016863713.1
XM_017008225.1 Intron XP_016863714.1
XM_017008226.1 Intron XP_016863715.1
XM_017008227.1 Intron XP_016863716.1
XM_017008228.1 Intron XP_016863717.1
XM_017008229.1 Intron XP_016863718.1
XM_017008230.1 Intron XP_016863719.1
XM_017008231.1 Intron XP_016863720.1
XM_017008232.1 Intron XP_016863721.1
XM_017008233.1 Intron XP_016863722.1
XM_017008234.1 Intron XP_016863723.1
XM_017008235.1 Intron XP_016863724.1

View Full Product Details