Product Details

SNP ID
rs79899235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:67618802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTGATCATAGCCACGTGTGAAC[A/C]GTTAGACAAGTGTATGCTATGCCCC
Phenotype
MIM: 611361
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UBA6 PubMed Links

Gene Details

Gene
UBA6
Gene Name
ubiquitin like modifier activating enzyme 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018227.5 3406 UTR 3 NP_060697.4
XM_017008359.1 3406 Intron XP_016863848.1

View Full Product Details