Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001134937.1 | Intron | NP_001128409.1 | |||
NM_001134938.1 | Intron | NP_001128410.1 | |||
NM_030917.3 | Intron | NP_112179.2 | |||
XM_005265769.4 | Intron | XP_005265826.1 | |||
XM_005265773.4 | Intron | XP_005265830.1 | |||
XM_005265774.4 | Intron | XP_005265831.1 | |||
XM_005265778.4 | Intron | XP_005265835.1 | |||
XM_005265779.4 | Intron | XP_005265836.1 | |||
XM_005265781.4 | Intron | XP_005265838.1 | |||
XM_005265782.4 | Intron | XP_005265839.1 | |||
XM_017008662.1 | Intron | XP_016864151.1 | |||
XM_017008663.1 | Intron | XP_016864152.1 | |||
XM_017008664.1 | Intron | XP_016864153.1 | |||
XM_017008665.1 | Intron | XP_016864154.1 | |||
XM_017008666.1 | Intron | XP_016864155.1 | |||
XM_017008667.1 | Intron | XP_016864156.1 | |||
XM_017008668.1 | Intron | XP_016864157.1 | |||
XM_017008669.1 | Intron | XP_016864158.1 | |||
XM_017008670.1 | Intron | XP_016864159.1 | |||
XM_017008671.1 | Intron | XP_016864160.1 |