Product Details
- SNP ID
-
rs77757018
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:53986199 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGAAGCTCAGTTTGGCTTCAGGGA[C/T]CATTTTAATTAAATTAATTATCTCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RPL21P44
PubMed Links
Gene Details
- Gene
- RPL21P44
- Gene Name
- ribosomal protein L21 pseudogene 44
There are no transcripts associated with this gene.
View Full Product Details