Product Details

SNP ID
rs75268290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177137485 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGTTGATTTGTATCTTAATTAT[G/T]CCTTGGTACTCCTTGGTTTCTTGGC
Phenotype
MIM: 606681
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NSD1 PubMed Links

Gene Details

Gene
NSD1
Gene Name
nuclear receptor binding SET domain protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022455.4 Intron NP_071900.2
NM_172349.2 Intron NP_758859.1

View Full Product Details