Product Details

SNP ID
rs77809587
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179552346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTGGTGCCTTCCAAGGAGCATC[A/G]TAGCACACACTCGAGGCTTCCTGAT
Phenotype
MIM: 610327
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
RUFY1 PubMed Links

Gene Details

Gene
RUFY1
Gene Name
RUN and FYVE domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040451.2 Intron NP_001035541.1
NM_001040452.2 Intron NP_001035542.1
NM_025158.4 Intron NP_079434.3
XM_005265993.4 Intron XP_005266050.1
XM_006714921.3 Intron XP_006714984.2
XM_006714922.3 Intron XP_006714985.1
XM_017009891.1 Intron XP_016865380.1
XM_017009892.1 Intron XP_016865381.1
XM_017009893.1 Intron XP_016865382.1
XM_017009894.1 Intron XP_016865383.1
XM_017009895.1 Intron XP_016865384.1

View Full Product Details