Product Details

SNP ID
rs76621265
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179733184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCGCGGTCATGGAGCGCCTTCG[C/T]CGGCGCATCGAGCTGTGCCGGCGCC
Phenotype
MIM: 114217 MIM: 605424
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CANX PubMed Links

Gene Details

Gene
CANX
Gene Name
calnexin
There are no transcripts associated with this gene.

Gene
MAML1
Gene Name
mastermind like transcriptional coactivator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014757.4 335 Silent Mutation CGC,CGT R24R NP_055572.1

View Full Product Details