Product Details
- SNP ID
-
rs77077795
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:180997241 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGCTCACTTCCTCTCATTTCCAT[C/T]GTGGGATATGTTGACGGAGGTATCC
- Phenotype
-
MIM: 606192
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BTNL3
PubMed Links
Gene Details
- Gene
- BTNL3
- Gene Name
- butyrophilin like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_197975.2 |
610 |
Silent Mutation |
ATC,ATT |
I142I |
NP_932079.1 |
View Full Product Details