Product Details

SNP ID
rs76427125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:161291842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAATGGCATTTTGGAGTTGTG[C/T]CCCTGAGTTATCAATATGTTCCTTC
Phenotype
MIM: 600232
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GABRB2 PubMed Links

Gene Details

Gene
GABRB2
Gene Name
gamma-aminobutyric acid type A receptor beta2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000813.2 3882 UTR 3 NP_000804.1
NM_021911.2 3882 UTR 3 NP_068711.1

View Full Product Details