Product Details

SNP ID
rs77781537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:30908134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGAACCAGTTCTCCTAATTGTA[A/G]CAAACAGTGCTGCAGAATCTGAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEDAG PubMed Links

Gene Details

Gene
MEDAG
Gene Name
mesenteric estrogen dependent adipogenesis
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032849.3 Intron NP_116238.2
XM_017020801.1 Intron XP_016876290.1
Gene
TEX26-AS1
Gene Name
TEX26 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details