Product Details

SNP ID
rs79868519
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:32303915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTAAAGCTGTAAATATTGCCACA[A/G]GAGAATCTCTTGTCTTTGCAGCGAC
Phenotype
MIM: 614818
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRY PubMed Links

Gene Details

Gene
FRY
Gene Name
FRY microtubule binding protein
There are no transcripts associated with this gene.

Gene
ZAR1L
Gene Name
zygote arrest 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136571.1 959 Silent Mutation TCC,TCT S310S NP_001130043.1

View Full Product Details