Product Details

SNP ID
rs76682711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:32305569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTAATACTCACATGGTGTTTAC[C/T]ATGTGCCTAGCACTCTCTTAAGCAC
Phenotype
MIM: 600185 MIM: 614818
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BRCA2 PubMed Links

Gene Details

Gene
BRCA2
Gene Name
BRCA2, DNA repair associated
There are no transcripts associated with this gene.

Gene
FRY
Gene Name
FRY microtubule binding protein
There are no transcripts associated with this gene.

Gene
ZAR1L
Gene Name
zygote arrest 1-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136571.1 Intron NP_001130043.1

View Full Product Details