Product Details

SNP ID
rs77645021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:23329359 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAATATGTACACACAAACATAA[A/G]GCAAGTGTTCTAACAGTTAATAAAT
Phenotype
MIM: 604490 MIM: 608896
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SACS PubMed Links

Gene Details

Gene
SACS
Gene Name
sacsin molecular chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278055.1 14601 UTR 3 NP_001264984.1
NM_014363.5 14601 UTR 3 NP_055178.3
XM_005266338.2 14601 UTR 3 XP_005266395.1
XM_011535039.2 14601 UTR 3 XP_011533341.1
XM_017020539.1 14601 UTR 3 XP_016876028.1
Gene
SGCG
Gene Name
sarcoglycan gamma
There are no transcripts associated with this gene.

View Full Product Details