Product Details

SNP ID
rs78279750
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:85793482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTATTTATAATTGTCAAGTTCCA[A/G]TTTCTTTTGCATTTATTGCAAGAAA
Phenotype
MIM: 609681
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
SLITRK6 PubMed Links

Gene Details

Gene
SLITRK6
Gene Name
SLIT and NTRK like family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032229.2 3486 UTR 3 NP_115605.2

View Full Product Details