Product Details

SNP ID
rs79624852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:85794605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTCTTTTTGTATCTTCTCCTG[T/C]GGTGAAGAACAAGAACCACTATCCC
Phenotype
MIM: 609681
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
SLITRK6 PubMed Links

Gene Details

Gene
SLITRK6
Gene Name
SLIT and NTRK like family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032229.2 2363 Missense Mutation CAC,CGC H635R NP_115605.2

View Full Product Details