Product Details

SNP ID
rs79517451
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:25247906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCACCATGTTATTTAAATTTTC[A/C]ATTTTAAAGCCAGTTGGGTGATATT
Phenotype
MIM: 603561
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
MTMR6 PubMed Links

Gene Details

Gene
MTMR6
Gene Name
myotubularin related protein 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004685.3 2776 UTR 3 NP_004676.3
XM_011535306.1 2776 UTR 3 XP_011533608.1
XM_011535307.1 2776 UTR 3 XP_011533609.1
XM_017020845.1 2776 UTR 3 XP_016876334.1
XM_017020846.1 2776 UTR 3 XP_016876335.1

View Full Product Details